Learn More
Cayman Chemical YTHDF2 PolycLNal ANTIBDY 500uL
SDP
Catalog No. 502937678
Encompass_Preferred
A flavonoid with anti-inflammatory and anxiolytic activities inhibits LPS-induced nitric oxide production in RAW 264 7 cells from 0 16-10 M reduces pulmonary edema and lung inflammation in a mouse model of LPS-induced acute lung injury at 10 mg/kg increases the number of entries and the time spent in the open arms of the elevated plus maze in rats
Catalog No. 50-293-7678 Supplier Cayman Chemical Supplier No. 27910500 UL
A flavonoid with anti-inflammatory and anxiolytic activities inhibits LPS-induced nitric oxide production in RAW 264 7 cells from 0 16-10 M reduces pulmonary edema and lung inflammation in a mouse model of LPS-induced acute lung injury at 10 mg/kg increases the number of entries and the time spent in the open arms of the elevated plus maze in rats