Learn More
Medchemexpress LLC Ytk-105 | 774192-20-0 | 98.8% | 257.33 g·mol⁻¹ | C16H19NO2 | 10 MG
SDP
Catalog No. 5000284195
Encompass_Preferred
YTK-105 is a small-molecule ligand of the p62 (SQSTM1) ZZ domain that activates p62-dependent selective macroautophagy and is used as a binder in AUTOTAC (autophagy-targeting chimera) design. Supplied as a high-purity research reagent, it is intended for in vitro and chemical biology applications and is not for human or clinical use.
  • Ligand of the p62-ZZ domain.
  • Activates p62-dependent selective macroautophagy.
  • Suitable for AUTOTAC design and targeted degradation studies.
  • High purity (>98.8%).
  • Supplied in small research quantities (for example, 10 mg).
Catalog No. 50-002-84195 Supplier Medchemexpress LLC Supplier No. HYW48912110MG
YTK-105 is a small-molecule ligand of the p62 (SQSTM1) ZZ domain that activates p62-dependent selective macroautophagy and is used as a binder in AUTOTAC (autophagy-targeting chimera) design. Supplied as a high-purity research reagent, it is intended for in vitro and chemical biology applications and is not for human or clinical use.
  • Ligand of the p62-ZZ domain.
  • Activates p62-dependent selective macroautophagy.
  • Suitable for AUTOTAC design and targeted degradation studies.
  • High purity (>98.8%).
  • Supplied in small research quantities (for example, 10 mg).