Learn More
Strem, An Ascensus Company CAS# 7440-64-4. 5g. Ytterbium chips (99.9% REO). MFCD00011286
SDP
Catalog No. 5090116232
Encompass
CAS# 7440-64-4. 5g. Ytterbium chips (99.9% REO). MFCD00011286. Molecular Weight: 173.04. Molecular Formula: Yb. Color/form: chips Strem# 93-7031. http://www.strem.com/catalog/v/93-7031/
Catalog No. 50-901-16232 Supplier Strem, An Ascensus Company Supplier No. 9370315G
Add to Cart
Add to Cart
CAS# 7440-64-4. 5g. Ytterbium chips (99.9% REO). MFCD00011286. Molecular Weight: 173.04. Molecular Formula: Yb. Color/form: chips Strem# 93-7031. http://www.strem.com/catalog/v/93-7031/