Learn More
Strem, An Ascensus Company CAS# 10361-91-8. 1g. Ytterbium(III) chloride, anhydrous (99.9%-Yb) (REO). MFCD00011288
SDP
Catalog No. 5090116225
Encompass
CAS# 10361-91-8. 1g. Ytterbium(III) chloride, anhydrous (99.9%-Yb) (REO). MFCD00011288. Molecular Weight: 279.40. Molecular Formula: YbCl3. Color/form: white pwdr. Strem# 93-7025. http://www.strem.com/catalog/v/93-7025/
Catalog No. 50-901-16225 Supplier Strem, An Ascensus Company Supplier No. 9370251G
CAS# 10361-91-8. 1g. Ytterbium(III) chloride, anhydrous (99.9%-Yb) (REO). MFCD00011288. Molecular Weight: 279.40. Molecular Formula: YbCl3. Color/form: white pwdr. Strem# 93-7025. http://www.strem.com/catalog/v/93-7025/