Learn More
Strem, An Ascensus Company CAS# 10035-01-5. 10g. Ytterbium(III) chloride hexahydrate (99.9%-Yb) (REO). MFCD00149885
SDP
Catalog No. 5090116215
Encompass
CAS# 10035-01-5. 10g. Ytterbium(III) chloride hexahydrate (99.9%-Yb) (REO). MFCD00149885. Molecular Weight: 279.40 (387.49). Molecular Formula: YbCl3∙6H2O. Color/form: white xtl. Strem# 93-7003. http://www.strem.com/catalog/v/93-7003/
Catalog No. 50-901-16215 Supplier Strem, An Ascensus Company Supplier No. 93700310G
CAS# 10035-01-5. 10g. Ytterbium(III) chloride hexahydrate (99.9%-Yb) (REO). MFCD00149885. Molecular Weight: 279.40 (387.49). Molecular Formula: YbCl3∙6H2O. Color/form: white xtl. Strem# 93-7003. http://www.strem.com/catalog/v/93-7003/