Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) chloride, ultra dry, 99.99% (REO)

Catalog No. AA4065314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4065314 25 g
AA4065306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4065314 Supplier Thermo Scientific Chemicals Supplier No. 04065314
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10361-91-8 | YbCl3

Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides. Ytterbium(III) chloride is a powerful catalyst for the formation of acetals using trimethyl orthoformate and also used to track digestion in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-91-8
Molecular Formula YbCl3
MDL Number MFCD00011288

Specifications

Melting Point 854°C
Color White
Physical Form Beads
Quantity 25 g
Odor Odorless
Sensitivity Hygroscopic
Assay Percent Range 99.99% (REO)
Solubility Information Slightly soluble in water.
Formula Weight 279.4
Chemical Name or Material Ytterbium(III) chloride, ultra dry
Hazard Category H315-H319-H335-H500
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-800-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.