Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) oxide, 99.9% (REO)

Catalog No. AA3625214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3625214 25 g
AA3625222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3625214 Supplier Thermo Scientific Chemicals Supplier No. 03625214
  • $242.00
    $237.65 / Each of 1

    Save $4.35

    2% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

CAS: 1314-37-0 | O3Yb2 | 394.09 g/mol

Ytterbium(III) oxide is being applied to numerous fiber amplifier and fiber optic technologies. High purity Ytterbium Oxide are widely applied as a doping agent for garnet crystals in lasers a important colorant in glasses and porcelain enamel glazes.. As Ytterbium Oxide has a significantly higher emissivity in the infrared range a higher radiant intensity is obtained with Ytterbium-based payloads.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-37-0
Molecular Formula O3Yb2
Molecular Weight (g/mol) 394.09
MDL Number MFCD00011290
InChI Key UZLYXNNZYFBAQO-UHFFFAOYSA-N
IUPAC Name diytterbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Yb+3].[Yb+3]

Specifications

Melting Point 2227°C
Physical Form Sintered Lump
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water,moderately soluble in strong mineral acids.
Formula Weight 394.08
Density 9.2 g/mL
Chemical Name or Material Ytterbium oxide
EINECSNumber 215-234-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.