Learn More
Ytterbium(III) oxide, 99.9% (REO)
Description
Ytterbium(III) oxide is being applied to numerous fiber amplifier and fiber optic technologies. High purity Ytterbium Oxide are widely applied as a doping agent for garnet crystals in lasers a important colorant in glasses and porcelain enamel glazes.. As Ytterbium Oxide has a significantly higher emissivity in the infrared range a higher radiant intensity is obtained with Ytterbium-based payloads.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2227°C |
| Physical Form | Sintered Lump |
| Quantity | 100 g |
| Assay Percent Range | 99.9% (REO) |
| Density | 9.2 |
| Solubility Information | Insoluble in water,moderately soluble in strong mineral acids. |
| Formula Weight | 394.08 |
| Chemical Name or Material | Ytterbium oxide |
Safety and Handling
| EINECSNumber | 215-234-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.