Learn More
Strem, An Ascensus Company CAS# 252976-51-5. 5g. Ytterbium(III) trifluoromethanesulfonate hydrate (Ytterbium triflate). MFCD03703499
SDP
Supplier:  Strem, An Ascensus Company 7060005G
Encompass
CAS# 252976-51-5. 5g. Ytterbium(III) trifluoromethanesulfonate hydrate (Ytterbium triflate). MFCD03703499. Molecular Weight: 620.25. Molecular Formula: Yb(SO3CF3)3∙XH2O. Color/form: white pwdr. Strem# 70-6000. http://www.strem.com/catalog/v/70-6000/
Catalog No. 50-901-12896