Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ytterbium(III) Trifluoromethanesulfonate

Catalog No. 4305955G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-059-55G 5 g
43-059-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-059-55G Supplier Sigma Aldrich Supplier No. 4305955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A water-tolerant Lewis acid catalyst recently used in one-pot syntheses of +a-lactams, and in stereocontrolled radical and nucleophilic addition reactions. Catalyzes the synthesis of deoxypenostatin A in a novel, stereoselective, intramolecular Diels-Alder reaction.Lewis acid catalyst used with a silylated chinchona alkaloid in an asymmetric synthesis of substituted +a-lactams from an acid chloride and aryl imine. Also used to catalyze an aldehyde-ene reaction in a synthesis of the guaiane skeleton.An important Lewis acid catalyst for a variety of synthetic reactions, including allylborations.

Specifications

CAS 54761-04-5
Molecular Formula C3F9O9S3Yb
Linear Formula (CF3SO3)3Yb
MDL Number MFCD06200261
Quantity 5 g
Synonym Yb(OTf)3; Trifluoromethanesulfonic acid ytterbium(III) salt; Yb(TFA)3; Ytterbium(III) triflate
Molecular Weight (g/mol) 620.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.