Learn More
AccuStandard, Inc YTTRIUM 100ML
SDP
Catalog No. NC2106832
Encompass
YTTRIUM 100ml | 10000 µg/mL in 2-5% Nitric Acid | 7440-65-5 | 100mL
This item is not returnable. View return policy
Catalog No. NC2106832 Supplier AccuStandard, Inc Supplier No. ICP69N10X1
YTTRIUM 100ml | 10000 µg/mL in 2-5% Nitric Acid | 7440-65-5 | 100mL