Learn More
SPEX CERTIPREP LLC 3P 1,000 ug/ml Yttrium Standard, 2% HNO3, 100ml
Catalog No. NC1149367
Encompass_Preferred
(HPS by Zeptometrix) 1,000 ug/ml Yttrium Standard, 2% HNO3, 100ml. The accuracy of all single-element standards is certified against NIST Spectrometric Standard Solutions where available. A Certificate of Analysis and Material Safety Data Sheet are included with each standard. All standards are certified accurate for a period of 18 months from the date of shipment unless noted otherwise.
Catalog No. NC1149367 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000671100
(HPS by Zeptometrix) 1,000 ug/ml Yttrium Standard, 2% HNO3, 100ml. The accuracy of all single-element standards is certified against NIST Spectrometric Standard Solutions where available. A Certificate of Analysis and Material Safety Data Sheet are included with each standard. All standards are certified accurate for a period of 18 months from the date of shipment unless noted otherwise.