Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Yttrium(III) butoxide solution

Catalog No. 51066125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-066-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-066-125ML Supplier Sigma Aldrich Supplier No. 51066125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Precursor to composite film electrode for solid oxide fuel cells.

Specifications

CAS 111941-71-0
Density 0.9 g/mL (at 25°C)
Boiling Point 109°C
Molecular Formula C12H27O3Y
Refractive Index n20/D 1.495
Linear Formula Y(OC4H9)3
Quantity 25 mL
MDL Number MFCD00797391
Molecular Weight (g/mol) 308.25
Percent Purity ≥99.9% trace metals basis
Biological Activity Central nervous system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.