Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Catalog No. AA1117814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1117814 25 g
AA1117822 100 g
AA1117836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1117814 Supplier Thermo Scientific Chemicals Supplier No. 01117814
Add to Cart

CAS: 13709-49-4 | F3Y | 145.90 g/mol

Yttrium(III) fluoride is used for the production of metallic yttrium, thin films, laboratory reagents, catalysts, electrical compounds, glasses and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-49-4
Molecular Formula F3Y
Molecular Weight (g/mol) 145.90
MDL Number MFCD00049714
InChI Key RBORBHYCVONNJH-UHFFFAOYSA-K
IUPAC Name yttrium(3+) trifluoride
SMILES [F-].[F-].[F-].[Y+3]

Specifications

Melting Point 1387°C
Physical Form Lump/Powder
Quantity 25 g
UN Number UN3288
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 145.9
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-257-5
RTECSNumber ZG3417500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.