Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Yttrium(III) nitrate hexahydrate

Catalog No. 23795725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-795-725G 25 g
23-795-7500G 500 g
23-795-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-795-725G Supplier Sigma Aldrich Supplier No. 23795725G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Source of yttrium used to make yttrium-based surfactant mesophases which are promising as adsorbing agents as well as for optically functional species.Hydration may vary slightly

Specifications

CAS 13494-98-9
Quantity 25 g
Molecular Formula N3O9Y -+ 6H2O
Linear Formula Y(NO3)3 -+ 6H2O
MDL Number MFCD00149940
Synonym Yttrium hexahydrate trinitrate
Molecular Weight (g/mol) 383.01
Percent Purity 99.8% trace metals basis
Density 2.682 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.