Learn More
Strem, An Ascensus Company CAS# 1314-36-9. 250g. Yttrium(III) oxide (99.99%-Y) (REO) PURATREM. MFCD00011473
SDP
Catalog No. 5090115208
Encompass
CAS# 1314-36-9. 250g. Yttrium(III) oxide (99.99%-Y) (REO) PURATREM. MFCD00011473. Molecular Weight: 225.81. Molecular Formula: Y2O3. Color/form: white pwdr. Strem# 93-3910. http://www.strem.com/catalog/v/93-3910/
Catalog No. 50-901-15208 Supplier Strem, An Ascensus Company Supplier No. 933910250G
CAS# 1314-36-9. 250g. Yttrium(III) oxide (99.99%-Y) (REO) PURATREM. MFCD00011473. Molecular Weight: 225.81. Molecular Formula: Y2O3. Color/form: white pwdr. Strem# 93-3910. http://www.strem.com/catalog/v/93-3910/