Learn More
Strem, An Ascensus Company CAS# 7446-33-5. 100g. Yttrium(III) sulfate octahydrate (99.9%-Y) (REO). MFCD00149946
SDP
Catalog No. 5090115213
Encompass
CAS# 7446-33-5. 100g. Yttrium(III) sulfate octahydrate (99.9%-Y) (REO). MFCD00149946. Molecular Weight: 465.99 (610.12). Molecular Formula: Y2(SO4)3∙8H2O. Color/form: white xtl. Strem# 93-3916. http://www.strem.com/catalog/v/93-3916/
Catalog No. 50-901-15213 Supplier Strem, An Ascensus Company Supplier No. 933916100G
CAS# 7446-33-5. 100g. Yttrium(III) sulfate octahydrate (99.9%-Y) (REO). MFCD00149946. Molecular Weight: 465.99 (610.12). Molecular Formula: Y2(SO4)3∙8H2O. Color/form: white xtl. Strem# 93-3916. http://www.strem.com/catalog/v/93-3916/