Learn More
Strem, An Ascensus Company CAS# 52093-30-8. 25g. Yttrium(III) trifluoromethanesulfonate, min. 98% (Yttrium triflate). MFCD00209623
SDP
Catalog No. 5090111525
Encompass
CAS# 52093-30-8. 25g. Yttrium(III) trifluoromethanesulfonate, min. 98% (Yttrium triflate). MFCD00209623. Molecular Weight: 536.12. Molecular Formula: Y(CF3SO3)3. Color/form: white pwdr. Strem# 39-4050. http://www.strem.com/catalog/v/39-4050/
Catalog No. 50-901-11525 Supplier Strem, An Ascensus Company Supplier No. 39405025G
CAS# 52093-30-8. 25g. Yttrium(III) trifluoromethanesulfonate, min. 98% (Yttrium triflate). MFCD00209623. Molecular Weight: 536.12. Molecular Formula: Y(CF3SO3)3. Color/form: white pwdr. Strem# 39-4050. http://www.strem.com/catalog/v/39-4050/