Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Yttrium(III) Tris(Isopropoxide)

Catalog No. 665916500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-591-6500MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 66-591-6500MG Supplier Sigma Aldrich Supplier No. 665916500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

In many asymmetric catalysis applications, glove box and Schlenk techniques should be employed to prevent exposure of the rare earth catalyst to air and moisture, which can be detrimental to the reaction outcome. Solutions of the catalyst should be made using anhydrous solvents and used shortly after preparation.Product of Kojundo Chemical Laboratory Co., Ltd

Specifications

CAS 99604-67-8
Molecular Formula C16H24N3PRuF6
Linear Formula C16H24N3PRuF6
MDL Number MFCD08457650
Quantity 250 mg
Synonym 2-Propanol yttrium(III) salt; Isopropyl alcohol yttrium(III) salt; Tris(isopropoxy) yttrium(III)
Molecular Weight (g/mol) 504.42 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.