Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium ingot, 99.9% (REO)

Catalog No. AA0039918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0039918 50 g
AA0039909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0039918 Supplier Thermo Scientific Chemicals Supplier No. 00039918
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-65-5 | Y | 88.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Quantity 50 g
Odor Odorless
Packaging Poly bottle
Form Ingot
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Contains less than 1% tantalum
Assay (REO)
Chemical Name or Material Yttrium ingot
EINECSNumber 231-174-8
TSCA Yes
storageNote1
  • Packaged in air stable poly bottle
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.