Learn More
SCP Science CONOSTAN 5000ppm Standard, Y 50g
Catalog No. NC9703443
Encompass
Single element, metallo-organic standards blended in 20 cSt blank oil. Complete with a Certificate of Analysis. For use with ICP, AA, rotrode, XRF, DCP, flame emission and other instruments. Custom blends are available and can be shipped within two working days.
Catalog No. NC9703443 Supplier SCP Science Supplier No. 150500395
Add to Cart
Add to Cart
Single element, metallo-organic standards blended in 20 cSt blank oil. Complete with a Certificate of Analysis. For use with ICP, AA, rotrode, XRF, DCP, flame emission and other instruments. Custom blends are available and can be shipped within two working days.