Learn More
Perkin Elmer US LLC Yttrium (Y) Pure Plus Standard, 10,000 µg/mL, 5% HNO3, 125 mL
Catalog No. 502111520
Encompass_Preferred
Concentration of Elements includes: Element Concentration Matrix Y 10,000 µg/mL 5% HNO3
Catalog No. 50-211-1520 Supplier Perkin Elmer US LLC Supplier No. N9304128
Concentration of Elements includes: Element Concentration Matrix Y 10,000 µg/mL 5% HNO3