Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ytx-465 | 2225824-53-1 | 99.1% | 458.51 | C25H26N6O3 | 50MG
SDP

Catalog No. 5000237735
Encompass_Preferred

YTX-465 is a small-molecule stearoyl-CoA desaturase inhibitor used in research applications, including studies of Parkinson's disease and other synucleinopathies. It is supplied as a white to off-white solid intended for in vitro research and biochemical assays, and demonstrates potent activity against the Ole1 target.

  • Inhibits Ole1 and SCD1 with IC50 values of 0.039 μM and 30.4 μM respectively.
  • Molecular weight 458.51 and formula C25H26N6O3.
  • High purity (≈99.1%) based on supplier assay results.
  • White to off-white solid suitable for formulation in DMSO.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple small pack sizes and as a 10 mM solution in DMSO for convenience.

Catalog No. 50-002-37735 Supplier Medchemexpress LLC Supplier No. HY12475150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YTX-465 is a small-molecule stearoyl-CoA desaturase inhibitor used in research applications, including studies of Parkinson's disease and other synucleinopathies. It is supplied as a white to off-white solid intended for in vitro research and biochemical assays, and demonstrates potent activity against the Ole1 target.

  • Inhibits Ole1 and SCD1 with IC50 values of 0.039 μM and 30.4 μM respectively.
  • Molecular weight 458.51 and formula C25H26N6O3.
  • High purity (≈99.1%) based on supplier assay results.
  • White to off-white solid suitable for formulation in DMSO.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple small pack sizes and as a 10 mM solution in DMSO for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.