Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YTX-465 | 2225824-53-1 | 99.1% | 458.51 | C25H26N6O3 | 5MG
SDP

Catalog No. 5000235798
Encompass_Preferred

YTX-465 is a research-grade small-molecule inhibitor of stearoyl-CoA desaturase (Ole1/SCD), supplied as a high-purity solid for biochemical and cell-based studies of lipid metabolism and related pathways.

  • Targets stearoyl-CoA desaturase (Ole1/SCD) with IC50 of 0.039 μM for Ole1 and 30.4 μM for SCD1.
  • Molecular formula C25H26N6O3 and molecular weight 458.51 g/mol.
  • High purity (99.1%) suitable for research applications.
  • Recommended storage: solid at 4°C protected from light; in solution -80°C for up to 6 months.
  • Available in multiple pack sizes and as a ready-to-use DMSO solution.
  • Provided for research use only; not for human or diagnostic use.

Catalog No. 50-002-35798 Supplier Medchemexpress LLC Supplier No. HY1247515MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YTX-465 is a research-grade small-molecule inhibitor of stearoyl-CoA desaturase (Ole1/SCD), supplied as a high-purity solid for biochemical and cell-based studies of lipid metabolism and related pathways.

  • Targets stearoyl-CoA desaturase (Ole1/SCD) with IC50 of 0.039 μM for Ole1 and 30.4 μM for SCD1.
  • Molecular formula C25H26N6O3 and molecular weight 458.51 g/mol.
  • High purity (99.1%) suitable for research applications.
  • Recommended storage: solid at 4°C protected from light; in solution -80°C for up to 6 months.
  • Available in multiple pack sizes and as a ready-to-use DMSO solution.
  • Provided for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.