Learn More
Medchemexpress LLC YTX-465 | 2225824-53-1 | 99.1% | 458.51 g·mol⁻1 | C25H26N6O3 | 100MG
SDP
Catalog No. 5000235834
Encompass_Preferred
YTX-465 is a research reagent that inhibits stearoyl-CoA desaturase (Ole1/SCD). It inhibits Ole1 (IC50 = 0.039 μM) and SCD1 (IC50 = 30.4 μM), and is supplied as high-purity material in solid form or as a DMSO solution for biochemical and cellular studies related to lipid metabolism and neurodegenerative disease research.
  • High purity (99.1%).
  • Inhibits Ole1 and SCD1 with reported IC50 values.
  • Available as solids and as DMSO solution for flexible dosing.
  • Provided with a product data sheet and a safety data sheet.
  • Molecular formula C25H26N6O3 and molecular weight 458.51 for reference.
Catalog No. 50-002-35834 Supplier Medchemexpress LLC Supplier No. HY124751100MG
YTX-465 is a research reagent that inhibits stearoyl-CoA desaturase (Ole1/SCD). It inhibits Ole1 (IC50 = 0.039 μM) and SCD1 (IC50 = 30.4 μM), and is supplied as high-purity material in solid form or as a DMSO solution for biochemical and cellular studies related to lipid metabolism and neurodegenerative disease research.
  • High purity (99.1%).
  • Inhibits Ole1 and SCD1 with reported IC50 values.
  • Available as solids and as DMSO solution for flexible dosing.
  • Provided with a product data sheet and a safety data sheet.
  • Molecular formula C25H26N6O3 and molecular weight 458.51 for reference.