Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YTX-465 | 2225824-53-1 | 99.1% | 458.51 g·mol⁻1 | C25H26N6O3 | 10MG
SDP

Catalog No. 5000235109
Encompass_Preferred

YTX-465 is a small-molecule inhibitor of stearoyl-CoA desaturase (SCD) developed for research use in studies of lipid metabolism and neurodegenerative disease models. It shows activity against yeast Ole1 and human SCD1 and is supplied as a high-purity research chemical for biochemical and cell-based assays.

  • Inhibits stearoyl-CoA desaturase (SCD) and yeast Ole1.
  • Reported IC50: 0.039 μM (Ole1) and 30.4 μM (SCD1).
  • High purity suitable for research applications.
  • Molecular formula C25H26N6O3 and molecular weight 458.51 g·mol⁻1.
  • Provided in milligram quantities for laboratory studies.

Catalog No. 50-002-35109 Supplier Medchemexpress LLC Supplier No. HY12475110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YTX-465 is a small-molecule inhibitor of stearoyl-CoA desaturase (SCD) developed for research use in studies of lipid metabolism and neurodegenerative disease models. It shows activity against yeast Ole1 and human SCD1 and is supplied as a high-purity research chemical for biochemical and cell-based assays.

  • Inhibits stearoyl-CoA desaturase (SCD) and yeast Ole1.
  • Reported IC50: 0.039 μM (Ole1) and 30.4 μM (SCD1).
  • High purity suitable for research applications.
  • Molecular formula C25H26N6O3 and molecular weight 458.51 g·mol⁻1.
  • Provided in milligram quantities for laboratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.