Learn More
Medchemexpress LLC Ytx-465 | 2225824-53-1 | 458.51 g·mol⁻1 | 25mg
SDP
Catalog No. 5000235267
Encompass_Preferred
YTX-465 is a stearoyl-CoA desaturase (Ole1/SCD) inhibitor YTX-465 inhibits Ole1 and SCD1 with IC50s of 0 039 M and 30 4 M respectively YTX-465 can be used in the research of Parkinson s disease and other synucleinopathies[1]
Catalog No. 50-002-35267 Supplier Medchemexpress LLC Supplier No. HY12475125MG
YTX-465 is a stearoyl-CoA desaturase (Ole1/SCD) inhibitor YTX-465 inhibits Ole1 and SCD1 with IC50s of 0 039 M and 30 4 M respectively YTX-465 can be used in the research of Parkinson s disease and other synucleinopathies[1]