Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Yu Fluorination Ligand

Catalog No. ALD005082 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD005082 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD005082 Supplier Sigma Aldrich Supplier No. ALD00508250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Yu Fluorination Ligand is widely employed for the chemoselective functionalization of sp2- and sp3 C-H bonds.The Yu Fluorination Ligand in tandem with Selectfluor™ (Aldrich 439479) under palladium catalysis can promote direct fluorination of C(sp3)-H bonds stereoselectively. Yu and coworkers have displayed this on amino acid derivatives with directing group assistance from the Yu-Wasa Auxiliary (Aldrich 791806).

Specifications

CAS 1622013-58-4
Molecular Formula C15H24N4O3
Linear Formula C15H24N4O3
Quantity 5 mg
Synonym tert-Butyl 4-(4-methyl-1H-pyrazole-1-carbonyl)-1,4-diazepane-1-carboxylate; SHI-16
Molecular Weight (g/mol) 308.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.