Learn More
Medchemexpress LLC HY-19977 5mg Medchemexpress, YU238259 CAS:1943733-16-1 Purity:>98%
SDP
Catalog No. 502082207
Encompass_Preferred
Medchemexpress, HY-19977 5mg YU238259 CAS:1943733-16-1 YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2207 Supplier Medchemexpress LLC Supplier No. HY199775MG
Medchemexpress, HY-19977 5mg YU238259 CAS:1943733-16-1 YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.