Learn More
Medchemexpress LLC Yuanhuacine (Gnidilatidin) | 60195-70-2 | 1 MG
SDP
Catalog No. 5000417678
Encompass_Preferred
Yuanhuacine (Gnidilatidin), a diterpene from *Daphne genkwa*, is an effective and highly selective inhibitor of the basal-like 2 (BL2) subtype of TNBC. It can induce G2/M cell cycle arrest and exhibits broad anti-tumor activity. Yuanhuacine is an orally active DNA damaging agent.
  • Highly selective inhibitor for the BL2 subtype of TNBC.
  • Induces G2/M cell cycle arrest.
  • Exhibits broad anti-tumor activity.
  • Acts as an orally active DNA damaging agent.
  • Inhibits proliferation in various cancer cell lines including H1993, H358, H460, Calu-1, H1299, A549, UMUC3, and HCT116.
  • Downregulates mTOR activation.
  • Inhibits migration of H1993 cells.
  • Upregulates p21 transcription and protein expression by regulating Sp1 protein stability.
Catalog No. 50-004-17678 Supplier Medchemexpress LLC Supplier No. HYN72251MG
Add to Cart
Add to Cart
Yuanhuacine (Gnidilatidin), a diterpene from *Daphne genkwa*, is an effective and highly selective inhibitor of the basal-like 2 (BL2) subtype of TNBC. It can induce G2/M cell cycle arrest and exhibits broad anti-tumor activity. Yuanhuacine is an orally active DNA damaging agent.
  • Highly selective inhibitor for the BL2 subtype of TNBC.
  • Induces G2/M cell cycle arrest.
  • Exhibits broad anti-tumor activity.
  • Acts as an orally active DNA damaging agent.
  • Inhibits proliferation in various cancer cell lines including H1993, H358, H460, Calu-1, H1299, A549, UMUC3, and HCT116.
  • Downregulates mTOR activation.
  • Inhibits migration of H1993 cells.
  • Upregulates p21 transcription and protein expression by regulating Sp1 protein stability.