Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Calcium AS

Catalog No. 23601549 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
1740 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-601-549 1740 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-549 Supplier HORIBA Supplier No. 1300047910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Yumizen C1200 Calcium AS used to test for Calcium

Reagents: 

  • R1:
    • Phosphate buffer
    • 8-Hydroxyquinoline-5-sulfonic acid
    • Arsenazo III

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Control Sets Four: N (Normal) Multi Control and P (Abnormal) Multi Control; Urine Control Level 1 and Urine Control Level 2
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Calcium
Sample Type Serum or Plasma
Sensitivity 0.48 mg/dL
Type Reagent
Disposable Single-use
For Use With (Application) Photometric Test using Arsenazo III
Packaging Type 3 x 4 x 4 in. Box
Quantity 1740 Tests
Species Human
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.