Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX SAS Yumizen C1200 Clinical Chemistry Analyzer

Catalog No. 23601539 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Sample Type:
Plasma, Serum, Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
23-601-539 Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 23-601-539 Supplier HORIBA ABX SAS Supplier No. 5300002210

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Next generation system designed to provide high efficiency and accuracy with optimal workflow capabilities for laboratories processing up to 2 million plus tests annually.

General Description:

  • Analysis method types: Spectrophotometry, colorimetric and immunoassay Flexible
  • Indirect ISE Na+, K+, and Cl- with reference electrode – reliable
  • Throughput 1200 tests per hour Efficient workflow for labor savings
  • Automatic maintenance during start-up and shutdown for labor savings

Sample Handling:

  • Sample tray holds 84 samples, continuous loading
  • Sample probes liquid level sensing, cap/shock and clot detection
  • Sample ID with Integrated barcode reader.
  • Sample types: Serum, plasma and urine
  • Sample collection format: 5, 7, 10 mL tubes and sample cups
  • ISE sample volume: 1-25 uL (0.1 uL increment)
  • Automatic sample dilution 1/2 to 1/200
  • Calibration and Control tray: 61 refrigerated positions for calibrators and controls

Reagent Handling:

  • Reagent tray: 2 trays with 45 reagent positions each
  • Refrigeration: all reagents refrigerated at 208 °C
  • Reagent Cartridge: 20 (with adaptors), 40 or 70 mL
  • Dispensing system: Two reagent  probes, 3-part reagent capability
  • Reagent ID: Integrated Barcode reader
  • Reagent volume: 5-300 uL

Reaction System:

  • Reaction tray: 231 reusable high quality plastic cuvettes
  • Reaction volume: 80 to 430 uL
  • Temperature Reaction: 37 °C +/- 0.1 with inert bath oil
  • Extended Reaction Time: 3, 4, 5, 10, 15 minutes
  • Photometer reaction cycle: 21 or 31 minutes
  • Homogenization  13.5 seconds
  • Automatic correction: Rotation and repeated left-right movements by stirrers, serum blank, cell blank and measurement point change

Optical System:

  • Light source:  Halogen lamp with cooling system
  • Principle wavelengths: Endpoint, rate reaction, 2 point rate, immunoassay 14 wavelengths (340-884 nm), mon and bichromatic

Physical specifications:

  • Analyzer Size: 122 W x 85 D x 110.8 H cm or 48 in. W x 33 in. D x 43 in. H
  • Analyzer weight: 450 kgs / 992 Lbs with AL 1208 lbs
  • Water Specifications: Conductivity < 1 us/cm

Specifications

Type Clinical Chemistry Analyzer
Sample Type Plasma, Serum, Urine
Description Na+, K+,Cl- and Reference electrode
For Use With (Application) Processing Chemistry Tests
Analysis Time 12 minutes for one CMP panel 29 minutes for 20 CMP samples
Sample Dilution Automatic sample dilution 1/2 to 1/202
Throughput 1200 Tests/hr.
Calibration Endpoint, rate reaction, 2 point rat, immunoassay
Analytical Method Spectrophotometry, Colorimetric and Turbidimetric
Capacity 84 samples continuous loading
Cuvettes Yes
Distilled Water Consumption 22 L/hr.
Format Floor Model
Certifications/Compliance CE Marked, FDA Cleared
Includes BOM List Analyzer, Computer, Monitor, Keyboard, Mouse, Power cords Printer Wash solutions Set of Electrodes ISE solutions and standards and USB key containing Daily guide, Operations manual
Reaction Vessel 233 reusable high quality plastic cuvettes
Read Length 3 to 15 minutes, measurement cycle of 13.5 seconds
Quantity 24
Shipping Condition Crates shipped on truck to hospital
Dimensions (D x W x H) 124 x 85 x 110.8 cm (WxDxH) / 48 x 33 x 43 in. (WxDxH)
Depth (English) 33 in.
Depth (Metric) 85 cm
Height (English) 43 in.
Height (Metric) 110.8 cm
Width (English) 48 in.
Width (Metric) 122 cm
Electrical Requirements 2.6 kVA, 220 V ± 10%, 50/60 Hz
Amperage 2.6 kVA
Frequency 50/60 Hz
Voltage 220 V
Data Management Features Computer with keyboard
External Computer Yes
Laser Features Yes
Operating Environment 18°C to 30°C 40% to 70% with no condensation
Operating System Windows Operating System
Quality Control Details Multi Cal, Protein Cal, Albumin U/CSF Cal, CRP Cl, RF Cal, Cystatin C Cal, Ferritin Cal, TPU Cal, HDL Cal, LDL Cal, hsCRP Cal, N multi, P multi, Protein L1,L2; Urine L1,L2;Albumin U/CSF L1,L2;CO2, Cystatin C L1,L2; Low CRPhs
ISE Type Indirect
Weight (English) 994 lb.
Weight (Metric) 451 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.