Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 CO2 Control

Catalog No. 23601596 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
3 Pk. 3 mL Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-601-596 3 Pk. 3 mL Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-596 Supplier HORIBA Supplier No. 1300023950

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Yumizen C1200 CO2 Control used to monitor CO2 assay

  • Used to monitor CO2 assay

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 25°C
Concentration Normal concentration
Detectable Analytes CO2
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Sample Type CO2
Technology Assay Control
Type Control
Disposable Single-use
Level Normal
Quantity 3 Pk. 3 mL Vials
Source aq. soln. contains sodium azide as preservative
Stability Stable for 3 months (at 2°C to 25°C)
Sterility Non-sterile
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.