Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 CO2

Catalog No. 23601546 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Format:
Ready to Use (RTU)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-601-546 Ready to Use (RTU)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-546 Supplier HORIBA Supplier No. 1300023833
Add to Cart
Add to Cart
This item is not returnable. View return policy

Yumizen C1200 CO2 used to test for CO2

Reagents: 

  • R1:
    • Buffer
    • PEP
    • PEPC
    • MDH
    • NADH

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Control Sets Two: N (Normal) Multi Control and P (Abnormal) Multi Control
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes CO2
Format Ready to Use (RTU)
Sample Type Serum or Plasma
Sensitivity Limit of Detection = 1.1 mmol/L
Type Reagent
Disposable Single-use
For Use With (Application) Enzymatic Test
No. of Reactions 960 Tests
Packaging Type 3 x 4 x 5 in. Box
Species Human
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.