Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Complement C4

Catalog No. 23601569 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Format:
Ready to Use (RTU)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-601-569 Ready to Use (RTU)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-569 Supplier HORIBA Supplier No. 1300023876
This item is not returnable. View return policy

Yumizen C1200 Complement C4 used to test for C4

Reagents:

  • R1:
    • TRIS
    • NaCl
  • R2:
    • TRIS
    • NaCl
    • Antihuman C4 antibody (goat)

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Control Sets Two: Protein Level 1 and Level 2 Controls
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Complement C4
Format Ready to Use (RTU)
Sample Type Serum or Plasma
Sensitivity Limit of Detection = 1 mg/dL
Type Reagent
Disposable Single-use
For Use With (Application) Immunoturbidimetric Test
Packaging Type 3 x 4 x 4 in. Box
Quantity 200
Species Human
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.