Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Cuvette Segments (21)

Catalog No. 23601626 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Yumizen C1200 Clinical Chemistry Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-601-626 Yumizen C1200 Clinical Chemistry Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-626 Supplier HORIBA Supplier No. 3200660169
Add to Cart
Add to Cart
This item is not returnable. View return policy

Yumizen C1200 Cuvette Segments (21) used for assay reactions

  • Used for assay reactions

Specifications

Description Cuvette segment with 21 cuvettes for Yumizen C1200
For Use With (Application) Used for Reaction of Samples and Reagents
For Use With (Equipment) Yumizen C1200 Clinical Chemistry Analyzer
Quantity 1 cuvette segment with 21 cuvettes
Height (English) 3 in.
Height (Metric) 7.62 cm
Width (English) 12 in.
Width (Metric) 30.48 cm
Voltage 0/1 V
Weight (English) 0.25 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.