Learn More
Description
- Used to calibrate ISE
Specifications
Specifications
| Type | Reagent |
| Description | ISE Internal Standard used to calibrate ISE |
| For Use With (Equipment) | Used on Yumizen C1200 Clinical Chemistry Analyzer |
| Quantity | 200 mL Bottle |
| Height (English) | 3 in. |
| Height (Metric) | 7.62 cm |
| Width (English) | 3 in. |
| Width (Metric) | 7.62 cm |
| Voltage | 0/1 V |
| Weight (English) | 6.76 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.