Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Multi Calibrator

Catalog No. 23601579 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Format:
Swabs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-601-579 Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-579 Supplier HORIBA Supplier No. 1300023890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Yumizen C1200 Multi Calibrator is used to calibrate several assays on Yumizen C1200 Clinical Chemistry Analyzer.

  • Used to calibrate protein assays on Yumizen C1200 Clinical Chemistry Analyzer
  • Lyophilized calibrator based on serum
  • Concentration and activities adjusted to ensure optimum calibration of HORIBA methods

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Concentration Normal for each analyte
Detectable Analytes Alkaline Phosphatase, Albumin, ALT, Amylase, AST, CK-NAC, Calcium, Cholesterol, BUN, Bilirubin Direct (DBIL), Creatinine, GGT, LDH, Lipase, Glucose, Iron, Lactic Acid, Magnesium, Bilirubin Total (TBIL), Phosphate, Total Protein, Triglycerides, UA, Urea
Format Swabs
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Sample Type Serum
Type Calibrator
Disposable Single-use
Level Reference
Quantity 6 Pk. 3 mL Vials
Source Porcine, Bovine, Rabbit, Human, Chicken
Stability 2 days (at 2°C to 8°C, except for DBil = 8 hours and Tbil = 1 day)
Sterility Non-sterile
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.