Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 P Multi Control

Catalog No. 23601589 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
6 Pk. 5 mL Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-601-589 6 Pk. 5 mL Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-589 Supplier HORIBA Supplier No. 1300023940
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Yumizen C1200 P Multi Control is an abnormal control used to monitor several assays on Yumizen C1200 Clinical Chemistry Analyzer.

  • Used to monitor several assays on Yumizen C1200 Clinical Chemistry Analyzer

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Concentration Normal concentration for analytes
Detectable Analytes Alkaline Phosphatase, ALT, Amylase, AST, Albumin, CK-NAC, GGT, Bilirubin Direct (DBIL), Calcium, Cholesterol, Creatinine, Glucose, BUN, LDH, Lipase, Iron, Lactic Acid, Magnesium, Phosphate, Bilirubin Total (TBIL), Total Protein, Triglycerides, Urea, UA
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Sample Type Serum
Type Control
Disposable Single-use
Level Abnormal
Quantity 6 Pk. 5 mL Vials
Source Human Blood Material (Serum) and Animal Origin Material
Stability After reconstitution stable for 7 days (at 2°C to 8°C)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.