Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Protein Control

Catalog No. 23601590 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Concentration:
Abnormal concentration for analytes
Normal concentration for analytes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23-601-590 Normal concentration for analytes
23-601-591 Abnormal concentration for analytes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-601-590 Supplier HORIBA Supplier No. 1300023944
Add to Cart
Add to Cart
This item is not returnable. View return policy

Yumizen C1200 Protein Control used to monitor protein assays

  • Used to monitor protein assays
  • Human plasma with assigned values determined by calibration with reference material

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Concentration Normal concentration for analytes
Detectable Analytes C3, C4, IgA, IgG, IgM, Transferrin (TRF)
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Sample Type Serum, Plasma
Type Control
Disposable Single-use
Quantity 3 Pk. 1 mL Vials
Source Based on Human Plasma
Stability Stable for 12 weeks (at 2°C to 8°C)
Sterility Non-sterile
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.