Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Sample cup (J cup)

Catalog No. 23601621 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
1000 Sample Cups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-601-621 1000 Sample Cups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-621 Supplier HORIBA Supplier No. 3200660119
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

Yumizen C1200 Sample cup (J cup) used for samples on the Yumizen C1200 Clinical Chemistry Analyzer

  • Used for samples on the Yumizen C1200 Clinical Chemistry Analyzer

Specifications

Description Sample cup J cup
For Use With (Application) Holds Serum or Plasma Samples on Analyzer
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Quantity 1000 Sample Cups
Height (English) 0.25 in.
Height (Metric) 6.35 mm
Width (English) 12 in.
Width (Metric) 30.48 cm
Voltage 0/1 V
Weight (English) 0.4 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.