Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Total Protein

Catalog No. 23601563 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Format:
Ready to Use (RTU)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-601-563 Ready to Use (RTU)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-563 Supplier HORIBA Supplier No. 1300023865
Add to Cart
Add to Cart
This item is not returnable. View return policy

Yumizen C1200 Total Protein used to test for Total Protein

Reagents:

  • R1:
    • Sodium hydroxide
    • Potassium sodium tartrate
  • R2:
    • Sodium hydroxide
    • Potasium sodium tartrate
    • Potassium iodide
    • Copper sulphate

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Control Sets Two: N (Normal) Multi Control and P (Abnormal) Multi Control
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Total Protein
Format Ready to Use (RTU)
Sample Type Serum or Plasma
Sensitivity Limit of Detection = 0.05 g/dL
Type Reagent
Disposable Single-use
For Use With (Application) Photometric Test using the Biuret Method
Packaging Type 3 x 4 x 5 in. Box
Quantity 1890
Species Human
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.