Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 TPU Calibrator

Catalog No. 23601587 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Quantity:
3 Pk. 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-601-587 3 Pk. 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-587 Supplier HORIBA Supplier No. 1300047913

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Yumizen C1200 TPU Calibrator used to calibrate urinary protein assay

  • Used to calibrate urinary protein assay

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Concentration Reference concentration
Detectable Analytes Urinary Proteins
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Sample Type Serum
Technology Assay Calibration
Type Calibrator
Disposable Single-use
Level Reference
Quantity 3 Pk. 3 mL
Source aq. soln. containing human serum
Stability 9 weeks (at 2°C to 8°C)
Sterility Non-sterile
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.