Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Urinary Proteins

Catalog No. 23601578 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Sample Type:
Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
23-601-578 Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-601-578 Supplier HORIBA Supplier No. 1300047912
This item is not returnable. View return policy

Yumizen C1200 Urinary Proteins used to test for urinary proteins

Reagents:

  • R1:
    • Pyrogallol red
    • Sodium molybdate
    • Detergents

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Control Sets Two: Urine Control Level 1 and Urine Control Level 2
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Urine Protein
Sample Type Urine
Sensitivity 1.21 mg/dL
Type Reagent
Disposable Single-use
For Use With (Application) Photometric Test using Pyrogallol Red-molybdate Complex
Packaging Type 3 x 4 x 4 in. Box
Quantity 930
Species Human
Stability 20 days on board refrigerated compartment
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.