Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen H2500 Hematology Analyzer (Package)

Catalog No. 22111011 Shop All ABX Diagnostics, Inc. Products
Change view
Click to view available options
Sample Type:
Body Fluid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
22-111-011 Body Fluid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-111-011 Supplier HORIBA Supplier No. 5300002225

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

Hematology Analyzer Package for slide preparation system

  • For use with Yumizen SPS

Specifications

Type Hematology Analyzer
Sample Type Body Fluid
Voltage 120 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.