Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ YUMM1.7 D4 Mouse Melanoma Cell Line

Catalog No. SCC227MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC227MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC227MI Supplier MilliporeSigma™ Supplier No. SCC227
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YUMM1.7D4 melanoma cell line is a diploid clone of YUMM1.7, developed as immunicompetent cell line

  • Immunocompetent cell line that recapitulates genetic drivers found in a significant proportion of human melanomas
  • Diploid genotype of YUMM1.7D4 cells greatly enhances efficiency of CRISPR-based screens with higher frequency of biallelic knockout compared to tetraploid cells
  • Syngeneic with the immunocompetent C57/B1/6 mouse background, retaining genetic markers of the Braf/Pten mouse model in which melanoma tumors develop with short latency

Contains genetically modified organisms (GMO)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cells can be cultured 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Cell Type Cancer Cells
For Use With (Application) Cell Culture
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.