Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ YUMM1.7 D5 Mouse Melanoma Cell Line SDS

Catalog No. SCC228MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC228MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC228MI Supplier MilliporeSigma™ Supplier No. SCC228
Add to Cart
Add to Cart

YUMM1.7D5 mouse melanoma cell line is a diploid clone of YUMM1.7 cell line developed as an immunicompetent cell line

  • Recapitulates genetic drivers found in a significant proportion of human melanomas
  • Originated from a 4-hydroxytamoxifen-induced melanoma tumor in a male C57/B1/6 mouse into which mutations from the Braf/Pten genetically-engineered mouse model had been introduced via backcrossing
  • Harbors Braf V600E mutation and is homozygous negative for wild-type Pten and Cdkn2
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cells can be cultured for 10 passages after thawing without significantly affecting cell marker expression and functionality
Cell Type Cancer Cells
For Use With (Application) Cell Culture
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.