Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ YUMM1.7 Mouse Melanoma Cell Line

Catalog No. SCC191MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC191MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC191MI Supplier MilliporeSigma™ Supplier No. SCC191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YUMM1.7 mouse melanoma cell line was developed as an immunocompetent cell line recapitulates genetic drivers found in a significant proportion of human melanomas.

  • Derived from a 4-hydroxytamoxifen-induced melanoma tumor in a male C57/B1/6 mouse into which mutations from the Braf/Pten genetically-engineered mouse model had been introduced via backcrossing

Contains genetically modified organisms (GMO)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cells cultured for at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Cell Type Cancer Cells
For Use With (Application) Cell Culture
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.