Learn More
ATCC YUMM1.7; Melanoma; Mouse (Mus musculus)
Catalog No. 502383570
Encompass
YUMM1.7 is a cell line with epithelial-like morphology isolated in 2014 from the skin of an adult male mouse with melanoma. This cell line is genetically modified containing human-relevant mutations like BrafV600E/wt (knock in conditionally activated Braf allele), Cdkn2a-/- Pten-/- (conditionally inactivated). It also has Tyrosinase:CreERT2 transgene, which restricts recombination to melanocytes. It was deposited by the Yale University School of Medicine.
Catalog No. 50-238-3570 Supplier ATCC Supplier No. CRL3362
YUMM1.7 is a cell line with epithelial-like morphology isolated in 2014 from the skin of an adult male mouse with melanoma. This cell line is genetically modified containing human-relevant mutations like BrafV600E/wt (knock in conditionally activated Braf allele), Cdkn2a-/- Pten-/- (conditionally inactivated). It also has Tyrosinase:CreERT2 transgene, which restricts recombination to melanocytes. It was deposited by the Yale University School of Medicine.