Learn More
ATCC YUMM4.1; Melanoma; Mouse (Mus musculus)
Catalog No. 502384107
Encompass
YUMM3.3 is a cell line with epithelial-like morphology isolated in 2015 from the skin of an adult female mouse with melanoma. This cell line is genetically modified containing human relevant mutations like Pten-/- (conditionally inactivated); Cdkn2a-/- (conditionally inactivated). It also has Tyrosinase:CreERT2 transgene which restricts recombination to melanocytes. It was deposited by the Yale University School of Medicine.
Catalog No. 50-238-4107 Supplier ATCC Supplier No. CRL3366
YUMM3.3 is a cell line with epithelial-like morphology isolated in 2015 from the skin of an adult female mouse with melanoma. This cell line is genetically modified containing human relevant mutations like Pten-/- (conditionally inactivated); Cdkn2a-/- (conditionally inactivated). It also has Tyrosinase:CreERT2 transgene which restricts recombination to melanocytes. It was deposited by the Yale University School of Medicine.